Assay Details
Target Gene Details
Entrez Gene ID: | 12995 |
Gene Name: | casein kinase 2, alpha 1 polypeptide |
Gene Aliases: |
Csnk2a1-rs4 |
Location: |
Chr.2:152226731-152281856 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Csnk2a1 | NM_007788.3 | 13 | 3236 | NP_031814.2 |
| AK042898.1 | 1 | 567 | ||
| AK162515.1 | 1 | 1311 | ||
| BC026149.1 | 13 | 3140 | AAH26149.1 | |
| BC060742.1 | 13 | 3191 | AAH60742.1 | |
| CA322616.1 |

Genomic Map