Assay Details
Target Gene Details
Entrez Gene ID: | 56431 |
Gene Name: | destrin |
Gene Aliases: |
2610043P17Rik, ADF, AU042046, Dsn, corn1, sid23p |
Location: |
Chr.2:143915331-143943324 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dstn | NM_019771.2 | 1 | 56 | NP_062745.1 |
| BF123977.1 | 1 | 67 |
More Information
Set Membership: |
|

Genomic Map