Assay Details
Target Gene Details
Entrez Gene ID: | 16500 |
Gene Name: | potassium voltage gated channel, Shab-related subfamily, member 1 |
Gene Aliases: |
Kcr1-1, Kv2.1, Shab |
Location: |
Chr.2:167095969-167188818 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnb1 | NM_008420.4 | 1 | 400 | NP_032446.2 |
| BC031776.1 | 1 | 356 | AAH31776.1 | |
| BC051422.1 | 1 | 357 | AAH51422.1 | |
| BC061501.1 | 1 | 381 | AAH61501.1 | |
| DQ646377.1 | 1 | 400 | ||
| EF437378.1 | 1 | 190 | ||
| M64228.1 | 1 | 205 | AAA40112.1 |
More Information
Set Membership: |
|

Genomic Map