Assay Details
Target Gene Details
Entrez Gene ID: | 16500 |
Gene Name: | potassium voltage gated channel, Shab-related subfamily, member 1 |
Gene Aliases: |
Kcr1-1, Kv2.1, Shab |
Location: |
Chr.2:167095969-167188818 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnb1 | NM_008420.4 | 2 | 3244 | NP_032446.2 |
| BC031776.1 | 2 | 3200 | AAH31776.1 | |
| BC061501.1 | 2 | 3225 | AAH61501.1 |
More Information
Set Membership: |
|

Genomic Map