Assay Details
Target Gene Details
Entrez Gene ID: | 241794 |
Gene Name: | potassium voltage-gated channel, subfamily G, member 1 |
Gene Aliases: |
AW536275, OTTMUSG00000016048 |
Location: |
Chr.2:168260110-168283440 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcng1 | NM_001081134.1 | 2 | 2076 | NP_001074603.1 |
More Information
Set Membership: |
|

Genomic Map