Assay Details
Target Gene Details
Entrez Gene ID: | 104362 |
Gene Name: | meiosis expressed gene 1 |
Gene Aliases: |
MLZ-278, Meg1 |
Location: |
Chr.2:3409043-3422648 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Meig1 | NM_008579.4 | 1 | 215 | NP_032605.1 |
| BC049538.1 | 1 | 215 | AAH49538.1 |

Genomic Map