Assay Details
Target Gene Details
Entrez Gene ID: | 100043899 |
Gene Name: | R3H domain containing-like |
Gene Aliases: |
OTTMUSG00000001070 |
Location: |
Chr.2:163491214-163504364 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| R3hdml | NM_001099331.2 | 1 | 357 | NP_001092801.1 |
More Information
Set Membership: |
|

Genomic Map