Assay Details
Target Gene Details
Entrez Gene ID: | 545474 |
Gene Name: | scratch homolog 2, zinc finger protein (Drosophila) |
Gene Aliases: |
AW060659 |
Location: |
Chr.2:152081529-152095802 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scrt2 | NM_001160410.1 | 1 | 550 | NP_001153882.1 |
More Information
Set Membership: |
|

Genomic Map