Assay Details
Target Gene Details
Entrez Gene ID: | 73024 |
Gene Name: | ER membrane protein complex subunit 7 |
Gene Aliases: |
2900064A13Rik, AI451465, ORF3, c11orf3 |
Location: |
Chr.2:112455014-112472155 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Emc7 | NM_133749.2 | 1 | 48 | NP_598510.1 |
| AJ250345.1 | CAC16213.1 | |||
| AJ278128.1 | CAC01616.1 | |||
| AK013744.1 | ||||
| AK088738.1 | BAC40540.1 | |||
| AK182314.1 | ||||
| BC115477.1 | ||||
| BC115478.1 |

Genomic Map