Assay Details
Target Gene Details
Entrez Gene ID: | 70827 |
Gene Name: | trafficking protein, kinesin binding 2 |
Gene Aliases: |
2900022D04Rik, 4733401O11Rik, AI480836, AW046271, Als2cr3, CALS-C, GRIF-1, GRIF1, OIP98, mKIAA0549 |
Location: |
Chr.1:58900449-58973482 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trak2 | NM_172406.3 | 13 | 1989 | NP_765994.2 |
| AK036650.1 | 13 | 1938 | BAC29517.1 | |
| AK122305.1 | 3 | 378 | BAC65587.1 | |
| BC060681.1 | 12 | 1706 | AAH60681.1 |
More Information
Set Membership: |
|

Genomic Map