Assay Details
Target Gene Details
Entrez Gene ID: | 70827 |
Gene Name: | trafficking protein, kinesin binding 2 |
Gene Aliases: |
2900022D04Rik, 4733401O11Rik, AI480836, AW046271, Als2cr3, CALS-C, GRIF-1, GRIF1, OIP98, mKIAA0549 |
Location: |
Chr.1:58900449-58973482 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trak2 | NM_172406.3 | 2 | 488 | NP_765994.2 |
| AK014647.1 | 2 | 438 | ||
| AK036650.1 | 2 | 437 | BAC29517.1 | |
| AK048261.1 | 2 | 437 | ||
| BC060681.1 | 1 | 205 | AAH60681.1 | |
| CJ116609.1 |
More Information
Set Membership: |
|

Genomic Map