Assay Details
Target Gene Details
Entrez Gene ID: | 18549 |
Gene Name: | proprotein convertase subtilisin/kexin type 2 |
Gene Aliases: |
6330411F23Rik, AI839700, Nec-2, Nec2, PC2, Phpp-2, SPC2 |
Location: |
Chr.2:143546133-143816284 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcsk2 | NM_008792.4 | 12 | 2773 | NP_032818.1 |
| BC057348.1 | 12 | 2728 | AAH57348.1 |
More Information
Set Membership: |
|

Genomic Map