Assay Details
Target Gene Details
Entrez Gene ID: | 74614 |
Gene Name: | osteoclast stimulatory transmembrane protein |
Gene Aliases: |
4833422F24Rik, OC-STAMP |
Location: |
Chr.2:165395450-165400394 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ocstamp | NM_029021.1 | 3 | 1246 | NP_083297.1 |
| AK014751.1 | 3 | 1246 | BAB29531.1 | |
| AK042271.1 | 3 | 1258 | BAC31210.1 | |
| BC020160.1 | 3 | 1214 | AAH20160.1 |

Genomic Map