Assay Details
Target Gene Details
Entrez Gene ID: | 76117 |
Gene Name: | Rho GTPase activating protein 15 |
Gene Aliases: |
5830480G12Rik |
Location: |
Chr.2:43748783-44395953 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgap15 | NM_153820.3 | 14 | 1987 | NP_722542.2 |
| AK036520.1 | ||||
| AK040796.1 | 15 | 2057 | ||
| BC024887.1 | 14 | 1941 | AAH24887.1 |
More Information
Set Membership: |
|

Genomic Map