Assay Details
Target Gene Details
Entrez Gene ID: | 13013 |
Gene Name: | cystatin 9 |
Gene Aliases: |
M12, cresp, testatin |
Location: |
Chr.2:148835147-148838737 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cst9 | NM_009979.1 | 1 | 94 | NP_034109.1 |
| AB017157.1 | 1 | 69 | BAA37089.1 | |
| AK006429.1 | 1 | 86 | BAB24584.1 | |
| BC048486.1 | 1 | 83 | AAH48486.1 | |
| Y18243.1 | 1 | 94 | CAA77090.1 |

Genomic Map