Assay Details
Target Gene Details
Entrez Gene ID: | 241303 |
Gene Name: | family with sequence similarity 78, member A |
Gene Aliases: |
A130092J06Rik |
Location: |
Chr.2:32052166-32087436 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam78a |
Target Gene Details
Entrez Gene ID: | 227720 |
Gene Name: | nucleoporin 214 |
Gene Aliases: |
BC039282, CAN, D2H9S46E |
Location: |
Chr.2:31974420-32053975 on Build GRCm38 |
Assay Gene Location: | Within Exon 36 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nup214 | NM_172268.2 | 36 | 6650 | NP_758472.2 |
| AK040284.1 | 8 | 2025 | ||
| BC039282.1 |
More Information
Set Membership: |
|

Genomic Map