Assay Details
Target Gene Details
Entrez Gene ID: | 20452 |
Gene Name: | ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 4 |
Gene Aliases: |
PST, PST-1, SIAT8-D, ST8SiaIV, Siat8d |
Location: |
Chr.1:95587682-95667649 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| St8sia4 | NM_001159745.1 | 1 | 228 | NP_001153217.1 |
| NM_009183.2 | 1 | 228 | NP_033209.1 | |
| AJ223956.1 | 1 | 223 | CAA11685.1 | |
| AK003690.1 | 1 | 170 | ||
| AK041723.1 | 1 | 161 | BAC31044.1 | |
| AK165717.1 | 1 | 203 | ||
| BC060112.1 | 1 | 205 | AAH60112.1 |
More Information
Set Membership: |
|

Genomic Map