Assay Details
Target Gene Details
Entrez Gene ID: | 20452 |
Gene Name: | ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 4 |
Gene Aliases: |
PST, PST-1, SIAT8-D, ST8SiaIV, Siat8d |
Location: |
Chr.1:95587682-95667649 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| St8sia4 | NM_001159745.1 | 3 | 788 | NP_001153217.1 |
| NM_009183.2 | 3 | 788 | NP_033209.1 | |
| AJ223956.1 | 3 | 779 | CAA11685.1 | |
| AK003690.1 | 3 | 730 | ||
| AK041723.1 | 3 | 721 | BAC31044.1 | |
| BC060112.1 | 3 | 765 | AAH60112.1 |
More Information
Set Membership: |
|

Genomic Map