Assay Details
Target Gene Details
Entrez Gene ID: | 20452 |
Gene Name: | ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 4 |
Gene Aliases: |
PST, PST-1, SIAT8-D, ST8SiaIV, Siat8d |
Location: |
Chr.1:95587682-95667649 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| St8sia4 | NM_009183.2 | 5 | 2568 | NP_033209.1 |
| AJ223956.1 | 5 | 2559 | CAA11685.1 | |
| AK041723.1 | 5 | 2501 | BAC31044.1 | |
| BC060112.1 | 5 | 2545 | AAH60112.1 |
More Information
Set Membership: |
|

Genomic Map