Assay Details
Target Gene Details
Entrez Gene ID: | 20846 |
Gene Name: | signal transducer and activator of transcription 1 |
Gene Aliases: |
2010005J02Rik, AA408197 |
Location: |
Chr.1:52119438-52161865 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stat1 | NM_001205313.1 | 25 | 4693 | NP_001192242.1 |
| NM_001205314.1 | 25 | 4711 | NP_001192243.1 | |
| NM_009283.4 | 25 | 4726 | NP_033309.3 | |
| BC042551.1 | 8 | 2858 | AAH42551.1 |
More Information
Set Membership: |
|

Genomic Map