Assay Details
Target Gene Details
Entrez Gene ID: | 66799 |
Gene Name: | ubiquitin-conjugating enzyme E2W (putative) |
Gene Aliases: |
6130401J04Rik |
Location: |
Chr.1:16540788-16619420 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ube2w | NM_001271016.1 | NP_001257945.1 | ||
| NM_001271017.1 | 6 | 1697 | NP_001257946.1 | |
| NM_025773.3 | 6 | 1776 | NP_080049.2 | |
| NR_073122.1 | 5 | 1673 | ||
| AK044463.1 | 6 | 1728 | BAC31935.1 | |
| AK050923.1 | BAE43345.1 | |||
| AK077997.1 | 6 | 1701 | BAC37094.1 | |
| AK082233.1 | 6 | 1725 | BAC38443.1 | |
| AK153165.1 | 1 | 1190 |
More Information
Set Membership: |
|

Genomic Map