Assay Details
Target Gene Details
Entrez Gene ID: | 12298 |
Gene Name: | calcium channel, voltage-dependent, beta 4 subunit |
Gene Aliases: |
3110038O15Rik, Cchb4, lh |
Location: |
Chr.2:52428320-52676609 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cacnb4 | NM_001037099.2 | 14 | 2652 | NP_001032176.1 |
| NM_001285426.1 | 14 | 2436 | NP_001272355.1 | |
| NM_001285427.1 | 14 | 2732 | NP_001272356.1 | |
| NM_001285428.1 | 13 | 2361 | NP_001272357.1 | |
| NM_146123.3 | 13 | 2572 | NP_666235.1 | |
| AF061330.1 | ||||
| AK047157.1 | 1 | 738 | ||
| AK147338.1 | 14 | 2626 | BAE27855.1 |
More Information
Set Membership: |
|

Genomic Map