Assay Details
Target Gene Details
Entrez Gene ID: | 66691 |
Gene Name: | GTPase activating protein and VPS9 domains 1 |
Gene Aliases: |
2010005B09Rik, 4432404J10Rik, AW108497, Gapex-5, RAP6, RME-6, mKIAA1521 |
Location: |
Chr.2:34676179-34755263 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gapvd1 | NM_025709.2 | 16 | 3152 | NP_079985.2 |
| AK014474.1 | BAB29377.2 | |||
| AK129381.1 | 15 | 2747 | BAC98191.1 | |
| AK134776.1 | 14 | 2142 | BAE22277.1 | |
| BC031478.1 | 13 | 2197 | ||
| BC048847.1 | 15 | 2718 | AAH48847.1 | |
| BC057164.1 | AAH57164.1 | |||
| BC060123.1 | 16 | 3152 | AAH60123.1 | |
| EF155419.1 | 14 | 2570 |
More Information
Set Membership: |
|

Genomic Map