Assay Details
Target Gene Details
Entrez Gene ID: | 19383 |
Gene Name: | hnRNP-associated with lethal yellow |
Gene Aliases: |
AI663842, Merc |
Location: |
Chr.2:154791097-154871010 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Raly | NM_001139511.1 | 2 | 481 | NP_001132983.1 |
| NM_001139512.1 | 3 | 247 | NP_001132984.1 | |
| NM_001139513.1 | 2 | 481 | NP_001132985.1 | |
| NM_023130.3 | 3 | 564 | NP_075619.2 | |
| AK014356.1 | 3 | 317 | BAB29294.1 | |
| AK135386.1 | 2 | 240 | BAE22515.1 | |
| AK152290.1 | 3 | 323 | ||
| AK154787.1 | 2 | 238 | BAE32830.1 | |
| AK168781.1 | 3 | 321 | BAE40615.1 | |
| AK185599.1 | 2 | 67 | ||
| AK215757.1 | 2 | 123 | ||
| BC004851.1 | 2 | 225 | AAH04851.1 | |
| BC016587.1 | 3 | 313 | AAH16587.1 | |
| BY028715.1 | 3 | 238 | ||
| CO812619.1 | 2 | 494 | ||
| CX214615.1 | 2 | 224 | ||
| L17076.1 | 3 | 294 | ||
| S72641.1 | 3 | 515 | AAC60688.1 |

Genomic Map