Assay Details
Target Gene Details
Entrez Gene ID: | 68428 |
Gene Name: | STEAP family member 3 |
Gene Aliases: |
1010001D01Rik, Tsap6, pHyde |
Location: |
Chr.1:120226414-120271082 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Steap3 | NM_001085409.1 | 3 | 678 | NP_001078878.1 |
| NM_133186.3 | 2 | 305 | NP_573449.2 | |
| AK028961.1 | 2 | 281 | ||
| AK078769.1 | 2 | 699 | BAC37383.1 | |
| AK167028.1 | 3 | 678 | BAE39201.1 | |
| AK171237.1 | 2 | 310 | BAE42333.1 | |
| AK203257.1 | 1 | 212 | ||
| AY029586.1 | 2 | 293 | AAK50539.1 | |
| BB857879.1 | 2 | 306 | ||
| BC037435.1 | 2 | 306 | AAH37435.1 | |
| BY718114.1 | 2 | 285 |

Genomic Map