Assay Details
Target Gene Details
Entrez Gene ID: | 68428 |
Gene Name: | STEAP family member 3 |
Gene Aliases: |
1010001D01Rik, Tsap6, pHyde |
Location: |
Chr.1:120226414-120271082 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Steap3 | NM_001085409.1 | 6 | 2877 | NP_001078878.1 |
| NM_133186.3 | 5 | 2504 | NP_573449.2 | |
| AK028961.1 | 5 | 2481 | ||
| AK078769.1 | BAC37383.1 | |||
| CD742498.1 | 1 | 266 |

Genomic Map