Assay Details
Target Gene Details
Entrez Gene ID: | 14563 |
Gene Name: | growth differentiation factor 5 |
Gene Aliases: |
BMP-14, Cdmp-1, bp, brp |
Location: |
Chr.2:155941023-155945364 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gdf5 | NM_008109.2 | 1 | 312 | NP_032135.2 |
| AB259648.1 | ||||
| BC034546.1 | 1 | 315 | AAH34546.1 | |
| BY015934.1 | 1 | 313 | ||
| U08337.1 | 1 | 315 | AAA18778.1 |
More Information
Set Membership: |
|

Genomic Map