Assay Details
Target Gene Details
Entrez Gene ID: | 67059 |
Gene Name: | Obg-like ATPase 1 |
Gene Aliases: |
2510025G09Rik, 2810405J23Rik, 2810409H07Rik, Gtpbp9, PTD004 |
Location: |
Chr.2:73092801-73214447 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ola1 | NM_025942.2 | 11 | 2018 | NP_080218.1 |
| AK013055.1 | 11 | 2019 | BAB28624.1 | |
| BX521012.1 | 1 | 232 |
More Information
Set Membership: |
|

Genomic Map