Assay Details
Target Gene Details
Entrez Gene ID: | 107746 |
Gene Name: | Rap guanine nucleotide exchange factor (GEF) 1 |
Gene Aliases: |
4932418O06Rik, C3G, Grf2 |
Location: |
Chr.2:29619720-29740364 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rapgef1 | NM_001039086.1 | 1 | 291 | NP_001034175.1 |
| NM_001039087.1 | 1 | 291 | NP_001034176.1 | |
| NM_054050.2 | 1 | 291 | NP_473391.1 | |
| AF348669.1 | 1 | 247 | AAK97373.1 | |
| AK147468.1 | 1 | 292 | BAE27934.1 | |
| AK147470.1 | 1 | 292 | BAE27936.1 | |
| AK147506.1 | 1 | 260 | BAE27960.1 | |
| AK147696.1 | 1 | 271 | BAE28079.1 | |
| BC129881.1 | 1 | 101 |
More Information
Set Membership: |
|

Genomic Map