Assay Details
Target Gene Details
Entrez Gene ID: | 19400 |
Gene Name: | receptor-associated protein of the synapse |
Gene Aliases: |
43kDa, Nraps, Raps, rapsyn |
Location: |
Chr.2:91035466-91045729 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rapsn | NM_009023.3 | 1 | 233 | NP_033049.2 |
| AK009672.1 | 1 | 234 | ||
| BC111863.1 | 1 | 189 | AAI11864.1 | |
| BY709325.1 | 1 | 234 | ||
| J03962.1 | 1 | 169 | AAA40030.1 | |
| X15788.1 | 1 | 222 | CAA33789.1 |

Genomic Map