Assay Details
Target Gene Details
Entrez Gene ID: | 19400 |
Gene Name: | receptor-associated protein of the synapse |
Gene Aliases: |
43kDa, Nraps, Raps, rapsyn |
Location: |
Chr.2:91035466-91045729 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rapsn | NM_009023.3 | 8 | 1386 | NP_033049.2 |
| AK009672.1 | 7 | 1230 | ||
| AK217703.1 | 2 | 172 | ||
| BC111863.1 | 8 | 1342 | AAI11864.1 | |
| J03962.1 | 8 | 1322 | AAA40030.1 | |
| X15788.1 | 8 | 1375 | CAA33789.1 |

Genomic Map