Assay Details
Target Gene Details
Entrez Gene ID: | 215653 |
Gene Name: | Ras association (RalGDS/AF-6) domain family member 2 |
Gene Aliases: |
3830431H01Rik, 9030412M04Rik, AI852669, AW495050 |
Location: |
Chr.2:131992850-132030287 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rassf2 | NM_175445.4 | 11 | 4192 | NP_780654.1 |
| AK018504.1 | 1 | 740 | ||
| AK172906.1 | 12 | 3968 | BAD32184.1 | |
| BC057402.1 | 11 | 4014 | AAH57402.1 |
More Information
Set Membership: |
|

Genomic Map