Assay Details
Target Gene Details
Entrez Gene ID: | 65964 |
Gene Name: | sterile alpha motif and leucine zipper containing kinase AZK |
Gene Aliases: |
AV006891, B230120H23Rik, MLTK, MLTKalpha, MLTKbeta |
Location: |
Chr.2:72285637-72442610 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zak | NM_001164791.1 | NP_001158263.1 | ||
| NM_023057.5 | NP_075544.1 | |||
| NM_178084.4 | 10 | 1233 | NP_835185.2 | |
| AB049731.1 | BAB16442.1 | |||
| AB049732.1 | BAB16443.1 | |||
| AK045444.1 | 10 | 1158 | BAC32371.1 | |
| AK087363.1 | ||||
| AK132186.1 | BAE21021.1 | |||
| AK162819.1 | BAE37068.1 | |||
| BC023718.1 | AAH23718.1 |
More Information
Set Membership: |
|

Genomic Map