Assay Details
Target Gene Details
Entrez Gene ID: | 227154 |
Gene Name: | STE20-related kinase adaptor beta |
Gene Aliases: |
AA792893, Als2cr2, B830008M19, D1Ucla2, ILPIP, ILPIPA, PRO1038, Papk, Syradb |
Location: |
Chr.1:58973527-58995126 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stradb | NM_172656.5 | 1 | 233 | NP_766244.4 |
| AB057666.1 | BAB68235.1 | |||
| AB057667.1 | BAB68236.1 | |||
| AK035207.1 | 1 | 164 | BAE43291.1 | |
| AK036678.1 | 1 | 228 | ||
| AK080096.1 | 1 | 197 | BAC37825.1 | |
| AK086974.1 | 1 | 164 | BAC39775.1 | |
| AK140272.1 | 1 | 283 | BAE24309.1 | |
| AK146079.1 | BAE26883.1 | |||
| BC060956.1 | 1 | 90 | AAH60956.1 | |
| CJ060534.1 | 1 | 283 |
More Information
Set Membership: |
|

Genomic Map