Assay Details
Target Gene Details
Entrez Gene ID: | 140579 |
Gene Name: | engulfment and cell motility 2 |
Gene Aliases: |
1190002F24Rik, CED-12 |
Location: |
Chr.2:165288031-165326479 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elmo2 | NM_080287.2 | 21 | 3744 | NP_525026.2 |
| NM_207706.1 | 22 | 3780 | NP_997589.1 | |
| AK035710.1 | 6 | 2130 | BAC29162.1 |
More Information
Set Membership: |
|

Genomic Map