Assay Details
Target Gene Details
Entrez Gene ID: | 241447 |
Gene Name: | ceramide synthase 6 |
Gene Aliases: |
4732462C07Rik, AW544719, Lass6, T1L |
Location: |
Chr.2:68861137-69114282 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cers6 | NM_172856.3 | 10 | 1546 | NP_766444.1 |
| AK028849.1 | 10 | 1547 | BAC26153.1 | |
| AK135798.1 | 10 | 1648 | ||
| AK151848.1 | BAE30739.1 | |||
| BC057629.1 | 10 | 1589 | AAH57629.1 |

Genomic Map