Assay Details
Target Gene Details
Entrez Gene ID: | 227624 |
Gene Name: | RAB, member RAS oncogene family-like 6 |
Gene Aliases: |
B230208H17Rik, Rbel1, Rbel1a, Rbel1b, parf |
Location: |
Chr.2:25583018-25608446 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rabl6 | NM_001024616.1 | 15 | 2517 | NP_001019787.2 |
| AK157020.1 | 15 | 2553 | ||
| AK162461.1 | 11 | 1897 | BAE36932.1 | |
| AK208962.1 | 1 | 171 | ||
| BC085507.1 | 15 | 2517 |
More Information
Set Membership: |
|

Genomic Map