Assay Details
Target Gene Details
Entrez Gene ID: | 56336 |
Gene Name: | UDP-Gal:betaGlcNAc beta 1,4-galactosyltransferase, polypeptide 5 |
Gene Aliases: |
9430078I07Rik, AW049941, AW539721 |
Location: |
Chr.2:167298444-167349178 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B4galt5 | NM_019835.2 | 9 | 3705 | NP_062809.2 |
| AK151469.1 | 6 | 3175 | BAE30427.1 | |
| AK152652.1 | 6 | 3175 | BAE31389.1 | |
| AK163380.1 | 1 | 1853 | ||
| AK190405.1 | 1 | 81 | ||
| AK191090.1 | 1 | 81 |
More Information
Set Membership: |
|

Genomic Map