Assay Details
Target Gene Details
Entrez Gene ID: | 54422 |
Gene Name: | BarH-like 1 (Drosophila) |
Gene Aliases: |
Dres115, MBH2 |
Location: |
Chr.2:28907680-28918415 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Barhl1 | NM_001164186.1 | 2 | 325 | NP_001157658.1 |
| NM_019446.4 | 1 | 608 | NP_062319.1 | |
| AB043980.1 | 1 | 623 | BAB18599.1 | |
| AF264026.1 | 3 | 383 | AAK58534.1 | |
| AK036312.1 | 1 | 59 | ||
| BC055731.1 | 2 | 325 | AAH55731.1 |
More Information
Set Membership: |
|

Genomic Map