Assay Details
Target Gene Details
Entrez Gene ID: | 21346 |
Gene Name: | transgelin 2 |
Gene Aliases: |
2700094C18Rik, SM22beta, Sm22B, Sm22a |
Location: |
Chr.1:172500246-172507375 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tagln2 | NM_178598.2 | 5 | 1107 | NP_848713.1 |
| AF149291.1 | 5 | 1098 | AAD37787.1 | |
| AF465519.1 | 6 | 1021 | AAM54133.1 | |
| AK019231.1 | 1 | 191 | ||
| AK151576.1 | 5 | 1107 | BAE30516.1 | |
| AK167395.1 | 5 | 1092 | BAE39484.1 | |
| AK167922.1 | 5 | 1089 | BAE39928.1 | |
| AK171088.1 | 6 | 1053 | BAE42241.1 | |
| AK172892.1 | 5 | 1046 | BAD32170.1 | |
| BC049861.1 | 5 | 1306 |
More Information
Set Membership: |
|

Genomic Map