Assay Details
Target Gene Details
Entrez Gene ID: | 68972 |
Gene Name: | TatD DNase domain containing 3 |
Gene Aliases: |
1500010M24Rik |
Location: |
Chr.1:191045826-191063010 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tatdn3 | NM_001163421.1 | 10 | 843 | NP_001156893.1 |
| NM_001163422.1 | 9 | 714 | NP_001156894.1 | |
| NM_026895.1 | 10 | 827 | NP_081171.1 | |
| AK005193.1 | 9 | 685 | BAB23875.1 | |
| AK156071.1 | 10 | 778 | BAE33572.1 | |
| BC061248.1 | 10 | 815 | AAH61248.1 |
More Information
Set Membership: |
|

Genomic Map