Assay Details
Target Gene Details
Entrez Gene ID: | 54610 |
Gene Name: | TBC1 domain family, member 8 |
Gene Aliases: |
AD3, HBLP1, VRP |
Location: |
Chr.1:39371495-39478779 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d8 | NM_018775.4 | 1 | 105 | NP_061245.3 |
| AK138630.1 | 1 | 103 | BAE23728.1 | |
| AK141520.1 | 1 | 94 | BAE24715.1 | |
| AK141816.1 | 1 | 59 | ||
| BC065081.1 | 1 | 39 | AAH65081.1 | |
| CJ139650.1 | 1 | 106 |
More Information
Set Membership: |
|

Genomic Map