Assay Details
Target Gene Details
Entrez Gene ID: | 66205 |
Gene Name: | CD302 antigen |
Gene Aliases: |
1110055L24Rik, AI159627 |
Location: |
Chr.2:60251993-60284484 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd302 | NM_001290660.1 | 1 | 246 | NP_001277589.1 |
| NM_025422.4 | 1 | 246 | NP_079698.2 | |
| AI116405.1 | 1 | 246 | ||
| AK004267.1 | 1 | 181 | BAB23242.2 |

Genomic Map