Assay Details
Target Gene Details
Entrez Gene ID: | 320706 |
Gene Name: | suppressor of glucose, autophagy associated 1 |
Gene Aliases: |
9830001H06Rik, AI426038, D430036N24Rik, Soga |
Location: |
Chr.2:157010442-157079265 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Soga1 | NM_001164663.1 | 1 | 524 | NP_001158135.1 |
| AK147227.1 | 1 | 514 |
Target Gene Details
Entrez Gene ID: | 383766 |
Gene Name: | TBC/LysM associated domain containing 2 |
Gene Aliases: |
Gm1332 |
Location: |
Chr.2:157075251-157096948 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tldc2 |

Genomic Map