Assay Details
Target Gene Details
Entrez Gene ID: | 227867 |
Gene Name: | enhancer of polycomb homolog 2 (Drosophila) |
Gene Aliases: |
5830499L14, D2Ertd694e |
Location: |
Chr.2:49451486-49551942 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Epc2 | NM_172663.4 | 13 | 2502 | NP_766251.3 |
| AK031025.1 | 13 | 2475 | BAC27218.1 | |
| AK047947.1 | 13 | 2170 | BAC33197.1 | |
| AK164886.1 | 9 | 1529 | BAE37953.1 | |
| BC048785.1 | 12 | 1941 | AAH48785.1 |
More Information
Set Membership: |
|

Genomic Map