Assay Details
Target Gene Details
Entrez Gene ID: | 17957 |
Gene Name: | N-ethylmaleimide sensitive fusion protein attachment protein beta |
Gene Aliases: |
Brp14, E161, I47, SNARE, b-SNAP |
Location: |
Chr.2:148693985-148732420 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Napb | NM_019632.3 | 11 | 3219 | NP_062606.1 |
| AK039204.1 | 1 | 217 | ||
| AK043917.1 | 1 | 279 | ||
| AK048037.1 | 1 | 305 | ||
| AK083184.1 | 11 | 3220 | BAC38798.1 | |
| BC016125.1 | 1 | 559 | ||
| BC038362.1 | 11 | 3193 | AAH38362.1 | |
| X61450.1 | 2 | 963 | CAA43690.1 |
More Information
Set Membership: |
|

Genomic Map