Assay Details
Target Gene Details
Entrez Gene ID: | 227721 |
Gene Name: | phosphatidic acid phosphatase type 2 domain containing 3 |
Gene Aliases: |
D830019K17Rik, NET39 |
Location: |
Chr.2:32095516-32110820 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppapdc3 | NM_145521.3 | 1 | 99 | NP_663496.1 |
| AK052883.1 | 1 | 100 | BAC35188.1 | |
| AK134638.1 | 1 | 223 | BAE22220.1 | |
| BC016136.1 | AAH16136.1 |
More Information
Set Membership: |
|

Genomic Map