Assay Details
Target Gene Details
Entrez Gene ID: | 19672 |
Gene Name: | reticulocalbin 1 |
Gene Aliases: |
Rcn |
Location: |
Chr.2:105385948-105399319 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rcn1 | NM_009037.2 | 1 | 295 | NP_033063.1 |
| AK017494.1 | 1 | 202 | BAB30773.1 | |
| AK133971.1 | 1 | 245 | BAE21962.1 | |
| AK159973.1 | 1 | 198 | BAE35525.1 | |
| BB612908.1 | ||||
| BC049108.1 | 1 | 188 | AAH49108.1 | |
| D13003.1 | 1 | 215 | BAA02366.1 |
More Information
Set Membership: |
|

Genomic Map