Assay Details
Target Gene Details
Entrez Gene ID: | 19700 |
Gene Name: | rad and gem related GTP binding protein 1 |
Gene Aliases: |
E030011C07Rik, Ras, Rem |
Location: |
Chr.2:152627008-152635191 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rem1 | NM_009047.5 | 2 | 386 | NP_033073.1 |
| AK053133.1 | 2 | 387 | BAC35274.1 | |
| AK084460.1 | 2 | 394 | BAC39190.1 | |
| BC150768.1 | 1 | 238 | ||
| U91601.1 | 2 | 364 | AAB71639.1 |

Genomic Map