Assay Details
Target Gene Details
Entrez Gene ID: | 228543 |
Gene Name: | ras homolog gene family, member V |
Gene Aliases: |
A030005A06Rik, Arhv |
Location: |
Chr.2:119269201-119278008 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rhov | NM_145530.2 | 3 | 1016 | NP_663505.1 |
| AK036775.1 | 3 | 1017 | BAC29571.1 | |
| AK037170.1 | 3 | 1018 | BAC29732.1 | |
| BC021307.1 | 3 | 1021 | AAH21307.1 |
More Information
Set Membership: |
|

Genomic Map